Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530291 Supplier Invitrogen™ Supplier No. PA530291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, HeLa nuclear extract, mouse brain, rat brain. Predicted reactivity: Mouse (99%), Rat (100%), Xenopus laevis (91%), Chicken (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the non-coding strand. This protein has been shown to function as an ATP-dependent DNA helicase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUBP1
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene FUBP1
Gene Accession No. Q32PX7, Q91WJ8, Q96AE4
Gene Alias 9530027K12Rik; D3Ertd330e; DNA helicase V; EMBL:AAI07943.1, ECO:0000312; EMBL:AAI07943.1}; far upstream element (FUSE) binding protein 1; far upstream element (FUSE) binding protein 4; far upstream element binding protein 1; far upstream element-binding protein; far upstream element-binding protein 1; far upstream element-binding protein 1 {ECO:0000250; FBP; FBP {ECO:0000250; Fubp; Fubp1; fubp1 {ECO:0000312; Fubp4; FUSE binding protein 1; FUSE-binding protein 1; FUSE-binding protein 1 {ECO:0000250; hDH V; RGD:1591892}; UniProtKB:Q96AE4, ECO:0000312; UniProtKB:Q96AE4}
Gene Symbols FUBP1
Host Species Rabbit
Immunogen Recombinant protein fragment corresponding to a region within amino acids 293 and 644 of Human FUBP1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51886, 654496, 8880
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.