Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUBP3 Monoclonal Antibody (1216CT820.155.150)
GREENER_CHOICE

Catalog No. PIMA537633
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537633 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537633 Supplier Invitrogen™ Supplier No. MA537633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FUBP3 Monoclonal Antibody for Western Blot, ICC/IF

May interact with single-stranded DNA from the far-upstream element (FUSE). May activate gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUBP3
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 1216CT820.155.150
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FUBP3
Gene Accession No. Q96I24
Gene Alias A330051M14Rik; AV006371; far upstream element (FUSE) binding protein 3; far upstream element binding protein 3; far upstream element-binding protein 3; FBP3; FUBP3; FUSE-binding protein 3; MAP2 RNA trans-acting protein 2; Marta2
Gene Symbols FUBP3
Host Species Mouse
Immunogen purified His-tagged FUBP3 protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.