Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUBP3 Polyclonal Antibody

FUBP3 Polyclonal Antibody
Supplier: Thermo Scientific PA596875
Description
FUBP3 Polyclonal Antibody for Western Blot, IHC (P)

Specifications
FUBP3 | |
Polyclonal | |
Unconjugated | |
FUBP3 | |
A330051M14Rik; AV006371; far upstream element (FUSE) binding protein 3; far upstream element binding protein 3; far upstream element-binding protein 3; FBP3; FUBP3; FUSE-binding protein 3; MAP2 RNA trans-acting protein 2; Marta2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
320267, 8939 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q96I24 | |
FUBP3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 65-260 of human FUBP3 (NP_0039251). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction