Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUCA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115256
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115256 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115256 Supplier Invitrogen™ Supplier No. PA5115256
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FUCA1.

Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids (Occhiodoro et al., 1989 [PubMed 2803312]). At least 2 separate polymorphic alpha-L-fucosidases are recognized in man: that in tissues, FUCA1, which is deficient in fucosidosis (MIM 230000), and that in plasma, FUCA2 (MIM 136820).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUCA1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Fuca1
Gene Accession No. P04066, P17164, Q99LJ1
Gene Alias 0610006A03Rik; 9530055J05Rik; A L fucosidase I; A L fucoside fucohydrolase 1; Afuc; alpha-L-fucosidase; alpha-L-fucosidase 1; Alpha-L-fucosidase I; alpha-L-fucoside fucohydrolase 1; EC 3.2.1; EC 3.2.1.51; Fuca; Fuca1; Fucosidase alpha-L-1 tissue; fucosidase, alpha-L- 1, tissue; Fucosidase, alpha-L-1, tissue; MGC137270 protein; Nbla10230; Tissue alpha-L-fucosidase; Tissue a L fucosidase; a L fucosidase 1; a L fucosidase I; a L fucoside fucohydrolase 1
Gene Symbols Fuca1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FUCA1(Accession P04066), corresponding to amino acid residues A121-A171.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24375, 2517, 71665
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.