Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUCA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584287
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584287 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584287 Supplier Invitrogen™ Supplier No. PA584287
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RDLVGELGTA LRKRNIRYGL YHSLLEWFHP LYLLDKKNGF KTQHFVSAKT MPELYDLVNS YKPDLIWSDG EWECPDTYWN STNFLSWL Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 90%.

Alpha-L-fucosidase (EC 3.2.1.51) is a lysosomal enzyme involved in the degradation of fucose-containing glycoproteins and glycolipids (Occhiodoro et al., 1989 [PubMed 2803312]). At least 2 separate polymorphic alpha-L-fucosidases are recognized in man: that in tissues, FUCA1, which is deficient in fucosidosis (MIM 230000), and that in plasma, FUCA2 (MIM 136820).[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUCA1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Fuca1
Gene Accession No. P04066
Gene Alias 0610006A03Rik; 9530055J05Rik; A L fucosidase I; A L fucoside fucohydrolase 1; Afuc; alpha-L-fucosidase; alpha-L-fucosidase 1; Alpha-L-fucosidase I; alpha-L-fucoside fucohydrolase 1; EC 3.2.1; EC 3.2.1.51; Fuca; Fuca1; Fucosidase alpha-L-1 tissue; fucosidase, alpha-L- 1, tissue; Fucosidase, alpha-L-1, tissue; MGC137270 protein; Nbla10230; Tissue alpha-L-fucosidase; Tissue α L fucosidase; α L fucosidase 1; α L fucosidase I; α L fucoside fucohydrolase 1
Gene Symbols Fuca1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FUCA1. Recombinant protein control fragment (Product #RP-105040).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2517
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.