Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUCA2 Monoclonal Antibody (234CT20.2.1)
GREENER_CHOICE

Catalog No. PIMA537516
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537516 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537516 Supplier Thermo Scientific Supplier No. MA537516
Add to Cart
Add to Cart

FUCA2 Monoclonal Antibody (234CT20.2.1)

FUCA2 Monoclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen FUCA2
Applications Western Blot
Classification Monoclonal
Clone 234CT20.2.1
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FUCA2
Gene Accession No. Q9BTY2
Gene Alias 0610025O11Rik; 5530401P20Rik; Alpha-L-fucosidase 2; alpha-L-fucoside fucohydrolase 2; dJ20N2.5; fc20c06; Fuca2; fucosidase, alpha-L- 2, plasma; MGC1314; plasma alpha-L-fucosidase; PSEC0151; UNQ227/PRO260; wu:fc20c06; zgc:92013
Gene Symbols FUCA2
Host Species Mouse
Immunogen FUCA2 recombinant protein.
Purification Method Protein G
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2519
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.