Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109741
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109741 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109741 Supplier Invitrogen™ Supplier No. PA5109741
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ADIFQASGAK YIVLTSKHHE GFTLWGSEYS WNWNAIDEGP KRDIVKELEV AIRNRTDLRF GLYYSLFEWF HPLFLEDESS SFHKRQFPVS KTLPELYELV NNYQPEVLWS DGDGGAPDQY WNSTGFLAWL YNESPVRGTV VTNDRWGAGS ICKHGGFYTC SDRYNPGHLL PHKWENCMTI DKLSWGYRRE AGISDYLTIE ELVKQLVETV SCGGNLLMNI GPTLDGTISV VFEERLRQMG SWLKVNGEAI YETHTWRSQN DTVTPDVWYT SKPKEKLVYA IFLKWPTSGQ LFLGHPKAIL GATEVKLLGH GQPLNWISLE QNGIMVELPQ LTIHQMPCKW GWALALTNVI.

FUCA2 (Alpha-L-fucosidase) catalyzes the hydrolysis of terminal alpha-L-fucosidase linkages in glycosphingolipids and glycoproteins. At least 2 separate polymorphic alpha-L-fucosidases are recognized in man. The FUCA2 locus regulates the level of alpha-fucosidase in plasma and fibroblasts but not in leukocytes. In fucosidosis, deficiency of alpha-L-fucosidase is found in both plasma and leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUCA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FUCA2
Gene Accession No. Q6AYS4, Q99KR8, Q9BTY2
Gene Alias 0610025O11Rik; 5530401P20Rik; Alpha-L-fucosidase 2; alpha-L-fucoside fucohydrolase 2; dJ20N2.5; fc20c06; Fuca2; fucosidase, alpha-L- 2, plasma; MGC1314; plasma alpha-L-fucosidase; PSEC0151; UNQ227/PRO260; wu:fc20c06; zgc:92013
Gene Symbols FUCA2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 118-467 of human FUCA2 (NP_114409.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2519, 292485, 66848
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.