Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUCA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112849
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112849 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112849 Supplier Invitrogen™ Supplier No. PA5112849
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUCA2 Polyclonal Antibody for Western Blot, IHC (P), ELISA

FUCA2 (Alpha-L-fucosidase) catalyzes the hydrolysis of terminal alpha-L-fucosidase linkages in glycosphingolipids and glycoproteins. At least 2 separate polymorphic alpha-L-fucosidases are recognized in man. The FUCA2 locus regulates the level of alpha-fucosidase in plasma and fibroblasts but not in leukocytes. In fucosidosis, deficiency of alpha-L-fucosidase is found in both plasma and leukocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUCA2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.75 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene FUCA2
Gene Accession No. Q9BTY2
Gene Alias 0610025O11Rik; 5530401P20Rik; Alpha-L-fucosidase 2; alpha-L-fucoside fucohydrolase 2; dJ20N2.5; fc20c06; Fuca2; fucosidase, alpha-L- 2, plasma; MGC1314; plasma alpha-L-fucosidase; PSEC0151; UNQ227/PRO260; wu:fc20c06; zgc:92013
Gene Symbols FUCA2
Host Species Rabbit
Immunogen Recombinant Human Plasma alpha-L-fucosidase protein (29-465AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2519
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.