Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fuchsin Solution, Decolorized R1, Ricca Chemical

Click to view available options
Quantity:
500 mL
1 L
Specifications
Specifications
Chemical Name or Material | FUCHSIN SOLUTION, DECOLORIZED R1 |
CAS | 7732-18-5, 7647-01-0, 569-61-9, 569-61-9 |
CAS Min % | 91.0, 6.46, 0.18, 0.05 |
CAS Max % | 95.0, 6.73, 0.19, 0.05 |
Grade | Laboratory |
Packaging | Amber Glass Bottle |
Color | Colorless to Yellow |
pH | <1 |
Quantity | 500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction