Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fucose Mutarotase Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300239APC
Description
Fucose Mutarotase Polyclonal antibody specifically detects Fucose Mutarotase in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
Fucose Mutarotase | |
Polyclonal | |
ELISA, Immunohistochemistry-Paraffin | |
C10orf125, chromosome 10 open reading frame 125, FucM, fucose mutarotase, FUCU | |
Produced in rabbits immunized with purified, recombinant Human Fucose Mutarotase (Accession#: A2VDF0-1; Met1-Leu154) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
282969 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction