Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fucose Mutarotase Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP300239UV
Description
Fucose Mutarotase Polyclonal specifically detects Fucose Mutarotase in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Fucose Mutarotase | |
Polyclonal | |
ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
C10orf125, chromosome 10 open reading frame 125, FucM, fucose mutarotase, FUCU | |
Produced in rabbits immunized with purified, recombinant Fucose Mutarotase (A2VDF0-1; Met1-Leu154). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
282969 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction