Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fucosyltransferase 3/FUT3 Mouse anti-Human, Guinea Pig, Clone: 2-25LE; same as LWB01, Novus Biologicals™

Mouse Monoclonal Antibody
$240.00 - $492.50
Specifications
Antigen | Fucosyltransferase 3/FUT3 |
---|---|
Clone | 2-25LE; same as LWB01 |
Concentration | 0.2 mg/ml |
Dilution | Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin) |
Description
Fucosyltransferase 3/FUT3 Monoclonal antibody specifically detects Fucosyltransferase 3/FUT3 in Human, Guinea Pig samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Fucosyltransferase 3/FUT3 | |
0.2 mg/ml | |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human, Guinea Pig | |
P21217 | |
2525 | |
IgG1 κ | |
Protein A or G purified |
2-25LE; same as LWB01 | |
Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
CD174, FT3B, FucT-III, LE, Les | |
Mucin isolated from a human ovarian cyst fluid (Uniprot: P21217) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title