Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fucosyltransferase 7/FUT7 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP302941100UL
Description
Fucosyltransferase 7/FUT7 Polyclonal specifically detects Fucosyltransferase 7/FUT7 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Fucosyltransferase 7/FUT7 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
2529 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
alpha-(1,3)-fucosyltransferase, EC 2.4.1, EC 2.4.1.-, EC 2.4.1.65, Fucosyltransferase 7, fucosyltransferase 7 (alpha (1,3) fucosyltransferase), Fucosyltransferase VII, fuc-TVII, FucT-VII, Galactoside 3-L-fucosyltransferase, Selectin ligand synthase, selectin-ligand synthase | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human Fucosyltransferase 7/FUT7 (NP_004470.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction