Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fuel Cell Car Science Kit

Click to view available options
Grade:
6+
Description
Includes
Car, solar panel, water-filling syringe, all necessary components, instruction manual and a Renewable Energy Education CD
- Uses a reversible PEM fuel cell that combines electrolysis and power conversion into one device
- Oxygen and hydrogen gases are formed in two transparent water containers
- Steers independently of the user once in operation
- Runs past obstacles; if the car hits a barrier, it automatically reverses 90°
- Grades: 6+
Renewable Energy Education CD
- Renewable Energy Science Education Manual: Introduces renewable energy in seven chapters—Global Warming and Climate Change, Solar Technologies, Wind Technologies, Electrolyzers, Fuel Cells, Hydrogen Storage, and Basic Electronics
- Experiment Manual: More than 20 experiments for middle to high school level sciences in several sections—Solar Energy, Wind Energy, Energy from Hydrogen (Fuel Cell), and “Ultra Cool« experiments
- Flash Animations: Describe and show how a fuel cell works, different types of fuel cells, electrolysis, and various applications for fuel cells
- Assembly Guides
- Technical Support with videos
Specifications
Specifications
Product Type | Fuel Cell Car Science Kit |
Grade | 6+ |
For Use With (Application) | Demonstrates electrolysis of water into oxygen and hydrogen |
For Use With (Equipment) | Electrolysis and power conversion into one single device |
Includes | Car, solar panel, water-filling syringe, all necessary components, instruction manual and a renewable energy education CD |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction