Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUK Monoclonal Antibody (6E2)
GREENER_CHOICE

Catalog No. PIMA515847
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515847 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515847 Supplier Invitrogen™ Supplier No. MA515847
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15847 targets FUK in indirect ELISA, FACS and WB applications and shows reactivity with Human, mouse, Non-human primate, and Rat samples. The MA5-15847 immunogen is purified recombinant fragment of human FUK expressed in E. Coli. . MA5-15847 detects FUK which has a predicted molecular weight of approximately 117kDa.

The protein encoded by this gene belongs to the GHMP (galacto-, homoserine, mevalonate and phosphomevalonate) kinase family and catalyzes the phosphorylation of L-fucose to form beta-L-fucose 1-phosphate. This enzyme catalyzes the first step in the utilization of free L-fucose in glycoprotein and glycolipid synthesis. L-fucose may be important in mediating a number of cell-cell interactions such as blood group antigen recognition, inflammation, and metastatis. While several transcript variants may exist for this gene, the full-length nature of only one has been described to date.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUK
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 6E2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene FUK
Gene Accession No. Q7TMC8, Q8N0W3
Gene Alias 1110046B12Rik; AI303278; Fcsk; Fucokinase; FUK; L-fucose kinase; mFLJ00241 protein; UNQ5838
Gene Symbols FUK
Host Species Mouse
Immunogen Purified recombinant human FUK (aa 622-795) expressed in E. coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 197258, 234730, 307848
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.