Learn More
Invitrogen™ FUK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120540
Description
Positive test controls include: U-87MG, Mouse brain, Mouse kidney. Immunogen sequence: LLKAAFICAG IVHVHSELQL SEQLLRTFGG GFELHTWSEL PHGSGLGTSS ILAGTALAAL QRAAGRVVGT EALIHAVLHL EQVLTTGGGW QDQVGGLMPG IKVGRSRAQL PLKVEVEEVT VPEGFVQKLN DHLLLVYTGK TRLARNLLQD VLRSWYARLP AVVQNAHSLV RQTEECAEGF RQGSLPLLGQ CLTSYWEQKK LMAPGCEPLT VRRMMDVLAP HVHGQSLAGA GGGGFLYLLT KEPQQKEALE AVLAKTEGLG NYSIHLVEVD TQGLSLKLLG TEASTCCPFP.
The protein encoded by this gene belongs to the GHMP (galacto-, homoserine, mevalonate and phosphomevalonate) kinase family and catalyzes the phosphorylation of L-fucose to form beta-L-fucose 1-phosphate. This enzyme catalyzes the first step in the utilization of free L-fucose in glycoprotein and glycolipid synthesis. L-fucose may be important in mediating a number of cell-cell interactions such as blood group antigen recognition, inflammation, and metastatis. While several transcript variants may exist for this gene, the full-length nature of only one has been described to date.
Specifications
| FUK | |
| Polyclonal | |
| Unconjugated | |
| FUK | |
| 1110046B12Rik; AI303278; Fcsk; Fucokinase; FUK; L-fucose kinase; mFLJ00241 protein; UNQ5838 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 197258, 234730 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.3 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7TMC8, Q8N0W3 | |
| FUK | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 795-1084 of human FUK (NP_659496.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.