Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ RU6500 Series Full-Facepiece Air-Purifying Respirators

Catalog No. 19085556 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Small
Medium
Large
Includes:
Head Strap
Head Strap and Mesh Head Net
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Includes
19-085-556 Medium Head Strap and Mesh Head Net
19-085-552 Small Head Strap
19-085-553 Medium Head Strap
19-085-554 Large Head Strap
19-085-555 Small Head Strap and Mesh Head Net
19-085-557 Large Head Strap and Mesh Head Net
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 19-085-556 Supplier Honeywell™ Supplier No. RU65002M
Add to Cart
Add to Cart
This item is not returnable. View return policy

Requires

Compatible with North™ N-Series cartridges, filters and accessories.

Features a polycarbonate lens with a wide field of vision for maximized peripheral vision plus a downward view.

  • Silicone seal provides secure facepiece seal, creating a barrier against contaminants
  • Silicone nose cup is soft and comfortable for all day wear
  • Wide viewing area provides distortion free visibility and optical clarity
  • Polycarbonate lens meet ANSI Z.87 for high impact
  • Facepiece with 5-point strap includes center strap to help position and hold the mask for a more secure fit; offers quick, easy adjustments
  • Facepiece with optional industrial nylon mesh head net (19-085-555, 556, 557) provides another choice for user comfort preference; can be used under hard hat
  • Threaded cartridge connector secures equipment to avoid accidental disconnections
  • Can be adapted to side- or front-mounted PAPR or CF-SAR; compatible with PA700 Series PAPR; PR500 Series Mask-Mounted PAPR; CF2000 Series SAR; and CF4000 Series Mask-Mounted SAR

Specifications

ANSI Standard ANSI Z87.1
Color Black
Material Silicone
Product Type Full Facepiece Respirator
Size Medium
Facepiece Material Silicone
For Use With (Equipment) North™ N-Series line of cartridges, filters, and cartridge/filter combinations
Includes Head Strap and Mesh Head Net
Material (Lens) Polycarbonate
Model RU65002M
Suspension Type 5-point
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.