Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell™ North™ 54001 Full Facepiece with Welding Attachment

Catalog No. 19085507 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-085-507 Medium/Large
19-085-508 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-085-507 Supplier Honeywell™ Supplier No. 54001W
Add to Cart
Add to Cart

Full facepiece elastomeric respirators with welding attachments feature a four strap headband, dual cartridge connectors for N series, a standard nose cup, polycarbonate lens, and a welding attachment. Cartridges, filters, parts, and accessories are sold separately.

  • Converts to PAPR or supplied air respirator
  • Lens offers 200 degree field of vision
  • Nose cup standard
  • Reduces fogging and increases worker comfort

Specifications

Color Black
Material Thermoplastic Elastomer
Product Type Full Facepiece Respirator with Welding Attachment
Size Medium/Large
Facepiece Material Thermoplastic Elastomer
For Use With (Equipment) North™ N-Series line of cartridges, filters, and cartridge/filter combinations
Includes Welding Attachment
Latex Free Yes
Model 54001W
Suspension Type 4-point
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.