Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Impacto™ Full Finger Anti-Fatigue Thermo Gloves
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Natural pain relief for prevention of injuries during work or play
- Designed to allow full ventilation during repetitive tasks, for extended periods of time
- Light but firm therapeutic compression counteracts tissue swelling to provide temporary relief from repetitive strain injuries (RSI)
- Promotes optimal muscle function at 1°C higher than the body’s normal resting temperature
- Ideal for repetitive jobs such as sewing, or for wearing inside other gloves when handling frozen products
Specifications
Specifications
Color | Black |
Material | Nylon |
Size | X-Small |
Finger Style | Full |
For Use With (Application) | Temporary Pain Relief from Repetative Strain Injuries (RSI) |
Glove Cuff Style | Hook and Loop |
Series | TS299 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction