Learn More
Invitrogen™ Fumarase Monoclonal Antibody (GT548)

Description
Positive Control: 293T, mouse liver, rat liver Predicted Reactivity: Zebrafish (90%), Pig (98%), Chicken (91%), Sheep (99%), Chimpanzee (99%), Bovine (98%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | Fumarase |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | GT548 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | FH |
| Gene Accession No. | P07954, P14408, P97807 |
| Gene Alias | EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.