Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fumarase Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596393
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596393 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596393 Supplier Invitrogen™ Supplier No. PA596393
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, LO2, 22Rv1, NCI-H460, HL-60, NIH/3T3, Mouse heart, Mouse kidney, Rat liver; Cellular Location: Cytoplasm, Mitochondrion Immunogen sequence: ASQNSFRIEY DTFGELKVPN DKYYGAQTVR STMNFKIGGV TERMPTPVIK AFGILKRAAA EVNQDYGLDP KIANAIMKAA DEVAEGKLND HFPLVVWQTG SGTQTNMNVN EVISNRAIEM LGGELGSKIP VHPNDHVNKS QSSNDTFPTA MHIAAAIEVH EVLLPGLQKL HDALDAKSKE FAQIIKIGRT HTQDAVPLTL GQEFSGYVQQ VKYAMTRIKA AMPRIYELAA GGTAVGTGLN TRIGFAEKVA AKVAALTGLP FVTAPNKFEA LAAHDALVEL SGAMNTTACS LMKIANDIRF LGSGPRSGLG ELILPENEPG SSIMPGKVNP TQCEAMTMVA AQVMGNHVAV TVGGSNGHFE LNVFKPMMIK NVLHSARLLG DASVSFTENC VVGIQANTER INKLMNESLM LVTALNPHIG YDKAAKIAKT AHKNGSTLKE TAIELGYLTA EQFDEWVKPK DMLGPK.

The protein encoded by this gene is an enzymatic component of the tricarboxylic acid cycle, or Krebs cycle, and catalyzes the formation of L-malate from fumarate. It exists in both a cytosolic form and an N-terminal extended form, differing only in the translation start site used. The N-terminal extended form is targeted to the mitochondrion, where the removal of the extension generates the same form as in the cytoplasm. It is similar to some thermostable class II fumarases and functions as a homotetramer. Mutations in this gene can cause fumarase deficiency and lead to progressive encephalopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fumarase
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene FH
Gene Accession No. P07954, P14408, P97807
Gene Alias EF-3; Fh; Fh1; Fh-1; FMRD; fumarase; fumarate hydratase; fumarate hydratase 1; fumarate hydratase, mitochondrial; HLRCC; LRCC; MCL; MCUL1
Gene Symbols FH, Fh1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 45-510 of human FH (NP_0001342).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14193, 14194, 2271, 24368
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.