Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Fumaric Acid
SCI_ED

Catalog No. s25333a Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g

Specifications

CAS 110-17-8
Color White
Density 1.625g/mL
Assay Percent Range >99% w/v
Packaging Solid
Molecular Formula C4H4O4
Linear Formula C4H4O4
Quantity 500 g
MDL Number MFCD00002700
Synonym fumaric acid, 2-butenedioic acid, trans-butenedioic acid, allomaleic acid, boletic acid, lichenic acid, tumaric acid, 2e-but-2-enedioic acid, butenedioic acid, trans-2-butenedioic acid
InChI Key VZCYOOQTPOCHFL-OWOJBTEDSA-N
SMILES OC(=O)\C=C\C(O)=O
IUPAC Name (2E)-but-2-enedioic acid
Molecular Weight (g/mol) 116.07
PubChem CID 444972
ChEBI CHEBI:18012
Formula Weight 116.07
Chemical Name or Material Fumaric Acid
Show More Show Less
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.