Learn More
Fumaric acid, 99%
Description
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Fumaric acid is used as a food additive and acid regulator. It is used in the production of polyester resins and polyhydric alcohols. It is also used as a mordant for dyes, paint additives, coating additives, plasticizers, processing aids and specific to petroleum production. Further, it is used in dentifrices as a stain remover and to make other chemicals. In addition to this, it is used in rosin esters, drying oils, printing inks and flavoring agent.
Solubility
Soluble in water, ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform, olive oil, carbon tetrachloride, xylene, molten camphor, liquid ammonia and benzene.
Notes
Incompatible with oxidizing agents, amines and strong bases.

Specifications
Specifications
Melting Point | ∼299°C (sublimation) |
Density | 1.625 |
pH | 2.1 |
Flash Point | 273°C (523°F) |
Odor | Odorless |
Quantity | 10,000 g |
Beilstein | 605763 |
Merck Index | 14,4287 |
Solubility Information | Soluble in water,ethanol and concentrated sulfuric acid. Slightly soluble in ethyl ether and acetone. Insoluble in chloroform,olive oil,carbon tetrachloride,xylene,molten camphor,liquid ammonia and benzene. |
Formula Weight | 116.07 |
Show More |
Safety and Handling
Hazard Category | H319-H500 |
Hazard Statement | GHS H Statement H319 Causes serious eye irritation. |
Precautionary Statement | P264b-P280i-P305+P351+P338 |
EINECSNumber | 203-743-0 |
RTECSNumber | LS9625000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.