Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fumonisin Monoclonal Antibody (2C7)

Catalog No. PIMA549616
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549616 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549616 Supplier Invitrogen™ Supplier No. MA549616

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

The fumonisins are a group of myco- toxins produced primarily by Fusarium verticillioides and Fusarium proliferatum. Maize is the most commonly contaminated crop. Fusarium verticillioides and fumonisins are found worldwide. The primary health concerns associated with fumonisins are acute toxic effects in horses and swine and carcinogenic properties. Fumonisins are carcinogenic to laboratory animals, and, in humans, consumption of fumonisin- contaminated maize is associated with higher rates of esophageal cancer and neural tube birth defects. Fumonisins were first characterized in the late 1980s, as a result of studies into the causes of esophageal cancer in Africa, coupled with outbreaks of equine leuko- encephalomalacia and porcine pulmonary edema in the U.S.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fumonisin
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 2C7
Concentration 4.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of FB hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.