Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUNDC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155388
Description
FUNDC1 Polyclonal specifically detects FUNDC1 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
FUNDC1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FUN14 domain containing 1, FUN14 domain-containing protein 1, MGC51029 | |
Rabbit | |
17 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunoprecipitation | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunoprecipitation | |
Q8IVP5 | |
FUNDC1 | |
Synthetic peptides corresponding to FUNDC1(FUN14 domain containing 1) The peptide sequence was selected from the N terminal of FUNDC1. Peptide sequence MATRNPPPQDYESDDDSYEVLDLTEYARRHQWWNRVFGHSSGPMVEKYSV. | |
Affinity purified | |
RUO | |
139341 | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction