Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUNDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548853
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548853 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548853 Supplier Invitrogen™ Supplier No. PA548853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with bovine, mouse and rat based on sequence homology.

Acts as an activator of hypoxia-induced mitophagy, an important mechanism for mitochondrial quality control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUNDC1
Applications Western Blot
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FUNDC1
Gene Accession No. Q8IVP5, Q9DB70
Gene Alias 1500005J14Rik; 1810033P05Rik; FUN14 domain containing 1; FUN14 domain-containing protein 1; fundc1; hypothetical protein LOC436984; si:ch211-222m21.2; zgc:92600
Gene Symbols FUNDC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 79-108 amino acids from the N-terminal region of human FUNDC1.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139341, 72018
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.