Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUNDC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558535
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558535 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA558535 Supplier Invitrogen™ Supplier No. PA558535
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MATRNPPPQD YESDDDSYEV LDLTEYARRH QWWNRVFGHS SGPMVEKYSV ATQIVM Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

Acts as an activator of hypoxia-induced mitophagy, an important mechanism for mitochondrial quality control.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUNDC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FUNDC1
Gene Accession No. Q8IVP5
Gene Alias 1500005J14Rik; 1810033P05Rik; FUN14 domain containing 1; FUN14 domain-containing protein 1; fundc1; hypothetical protein LOC436984; si:ch211-222m21.2; zgc:92600
Gene Symbols FUNDC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FUNDC1. Recombinant protein control fragment (Product #RP-95566).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 139341
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.