Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUNDC1 Recombinant Rabbit Monoclonal Antibody (8W4R3)
FUNDC1 Recombinant Rabbit Monoclonal Antibody (8W4R3)
Supplier: Thermo Scientific MA551458
Description
FUNDC1 Recombinant Monoclonal Antibody for IHC (P)

Specifications
FUNDC1 | |
Recombinant Monoclonal | |
0.29 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
Q5BJS4 | |
FUNDC1 | |
A synthesized peptide derived from human FUNDC1. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
Liquid |
ELISA, Immunohistochemistry (Paraffin) | |
8W4R3 | |
Unconjugated | |
FUNDC1 | |
1500005J14Rik; 1810033P05Rik; FUN14 domain containing 1; FUN14 domain-containing protein 1; fundc1; hypothetical protein LOC436984; si:ch211-222m21.2; zgc:92600 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
363442 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction