Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FUNDC2 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP231745

 View more versions of this product

Catalog No. NBP231745


Only null left
Add to Cart

Description

Description

FUNDC2 Polyclonal specifically detects FUNDC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

FUNDC2
Polyclonal
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Q9BWH2
FUNDC2
This antibody was developed against a recombinant protein corresponding to amino acids: MAASSQGNFEGDIESVDLAEFAKQQPWWRKLFGPESGLSAEKYSVATHLFIG
0.1 mL
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
cervical cancer oncogene 3, Cervical cancer proto-oncogene 3 protein, DC44, FLJ33773, FUN14 domain containing 2, FUN14 domain-containing protein 2, HCBP6MGC131676, HCC3, HCC-3, Hepatitis C virus core-binding protein 6, MGC2495, PD03104
Rabbit
Affinity Purified
RUO
65991
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only