Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FUNDC2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572155
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA572155 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA572155 Supplier Invitrogen™ Supplier No. PA572155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FUNDC2 Polyclonal Antibody for Western Blot

FUNDC2 (FUN14 Domain Containing 2) is a Protein Coding gene. An important paralog of this gene is FUNDC1. [GeneCards].
TRUSTED_SUSTAINABILITY

Specifications

Antigen FUNDC2
Applications Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% ProClin 300; pH 7.4
Gene Fundc2
Gene Accession No. Q9BWH2, Q9D6K8
Gene Alias 4833415N24Rik; cervical cancer oncogene 3; cervical cancer proto-oncogene 3 protein; DC44; FUN14 domain containing 2; FUN14 domain-containing protein 2; Fundc2; Hcbp6; HCBP6-like protein; HCC3; HCC-3; Hepatitis C virus core-binding protein 6; PD03104
Gene Symbols Fundc2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 122-150 amino acids from the C-terminal region of human FUNDC2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65991, 67391
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.