Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fungal Enolase Monoclonal Antibody (H67H)
GREENER_CHOICE

Catalog No. PIMA518201
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA518201 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA518201 Supplier Invitrogen™ Supplier No. MA518201
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with Candida species C. albicans, C. parapsilosis, C. tropicales and C. glabrata, as well as S. cerevisiae. Purity is >90%. Recommended pairs for ELISA: MA5-18202 or MA5-18203 (capture) and MA5-18201 (conjugated, detection).

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fungal Enolase
Applications ELISA
Classification Monoclonal
Clone H67H
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Host Species Mouse
Immunogen Enolase recombinant protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Fungi
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.