Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FUOM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551320
Description
The antibody detects endogenous levels of total FUOM protein.
Involved in the interconversion between alpha- and beta-L-fucoses. L-Fucose (6-deoxy-L-galactose) exists as alpha-L-fucose (29.5%) and beta-L-fucose (70.5%), the beta-form is metabolized through the salvage pathway. GDP-L-fucose formed either by the de novo or salvage pathways is transported into the endoplasmic reticulum, where it serves as a substrate for N- and O-glycosylations by fucosyltransferases. Fucosylated structures expressed on cell surfaces or secreted in biological fluids are believed to play a critical role in cell-cell adhesion and recognition processes.
Specifications
FUOM | |
Polyclonal | |
Unconjugated | |
FUOM | |
C10orf125; FucM; Fucose mutarotase; FUCU; FUOM; protein fucU homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
282969 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.2 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
A2VDF0 | |
FUOM | |
Full length fusion protein of human FUOM. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction