Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FUOM Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558696
Description
Immunogen sequence: MVALKGVPAL LSPELLYALA RMGHGDEIVL ADLNFPASSI CQCGPMEIRA DGLGIPQLLE AVL Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
Involved in the interconversion between alpha- and beta-L-fucoses. L-Fucose (6-deoxy-L-galactose) exists as alpha-L-fucose (29.5%) and beta-L-fucose (70.5%), the beta-form is metabolized through the salvage pathway. GDP-L-fucose formed either by the de novo or salvage pathways is transported into the endoplasmic reticulum, where it serves as a substrate for N- and O-glycosylations by fucosyltransferases. Fucosylated structures expressed on cell surfaces or secreted in biological fluids are believed to play a critical role in cell-cell adhesion and recognition processes.
Specifications
FUOM | |
Polyclonal | |
Unconjugated | |
FUOM | |
C10orf125; FucM; Fucose mutarotase; FUCU; FUOM; protein fucU homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
282969 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A2VDF0 | |
FUOM | |
Recombinant protein corresponding to Human FUOM. Recombinant protein control fragment (Product #RP-97109). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction