Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUR1 Antibody (OTI2D1), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272399AF750
Description
FUR1 Monoclonal specifically detects FUR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
FUR1 | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
UPRT | |
Full length recombinant protein of human FUR1 (NP_659489) produced in HEK293T cells. | |
DNA replication Transcription Translation and Splicing | |
139596 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI2D1 | |
Western Blot, Immunohistochemistry | |
DKFZp781E1243, EC 2.4.2, EC 2.4.2.9, FUR1, MGC23937, RP11-311P8.3, UMP pyrophosphorylase, UPP, UPRTase, uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae), uracil phosphoribosyltransferase homolog | |
Mouse | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction