Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Fura-2, Pentapotassium Salt, cell impermeant

Catalog No. F1200
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1200 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1200 Supplier Invitrogen™ Supplier No. F1200
Season of Savings
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Fura-2, pentapotassium salt is an intracellular calcium indicator that is ratiometric and UV light-excitable.

Fura-2, pentapotassium salt is an intracellular calcium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (Cat. No. I14402).

Fura-2, pentapotassium salt is an intracellular calcium indicator that is ratiometric and UV light-excitable. This water-soluble salt form is useful for intracellular loading by microinjection, infusion from patch pipette or uptake induced by our Influx pinocytic cell-loading reagent (Cat. No. I14402).

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in freezer (-5°C to -30°C) and protect from light.
Detection Method Fluorescence
For Use With (Application) Calcium Indicator
For Use With (Equipment) Fluorescence Microscope
Product Type Fura-2
Dye Type Fura-2
Excitation Wavelength Range 330 to 345 nm
Quantity 1 mg
Shipping Condition Room Temperature
Emission Wavelength Range 360 to 375 nm
Excitation Wavelength 339 nm
Emission Wavelength 364 nm
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.