Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ FURA-2AM
GREENER_CHOICESDP

Catalog No. 22201 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-201 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-201 Supplier R&D Systems™ Supplier No. 2220/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Fluorescent Ca2+ indicator

FURA-2AM is a fluorescent ratiometric Ca2+ indicator. FURA-2AM is selective for Ca2+ over other divalent cations Mg2+, Zn2+, Fe2+ and Mn2+. FURA-2AM binds to free intracellular calcium and is used to determine [Ca2+]i concentration. This product is typically prepared in DMSO. F 127 (Cat. No. 6253) for the solubilization of FURA-2AM is also available.

Chemical Identifiers

CAS 108964-32-5
Molecular Formula C44H47N3O24

Specifications

Quantity 1 mg
Product Line Tocris
Formula Weight 1001.85
Purity >95%
Molecular Weight (g/mol) 1001.857
Chemical Name or Material FURA-2AM
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.