Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Furamidine dihydrochloride
GREENER_CHOICE

Catalog No. 520225
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-022-5 25 mg
52-025 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-022-5 Supplier Tocris Bioscience™ Supplier No. 5202/25
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective PRMT1 inhibitor

Furamidine dihydrochloride is a selective protein arginine methyltransferase 1 (PRMT1) inhibitor (IC50 = 9.4 μM). Exhibits selectivity over PRMT5, PRMT6 and CARM1 (IC50 values are 166, 283 and >400 μM respectively). Inhibits cell proliferation in leukemia cell lines. Cell-permeable.

Specifications

Chemical Name or Material 4,4'-(2,5-Furandiyl)bis-benzenecarboximidamide dihydrochloride
CAS 55368-40-6
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C18H16N4O.2HCl
Formula Weight Observed MW: 377.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.