Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furan, 99+%, stabilized, ACROS Organics™

Click to view available options
Quantity:
250mL
1L
2.5L
Packaging:
Glass bottle
110-00-9,128-37-0
C4H4O
68.06
MFCD00003222
YLQBMQCUIZJEEH-UHFFFAOYSA-N
divinylene oxide,furfuran,tetrole,oxole,oxacyclopentadiene,furane,1,4-epoxy-1,3-butadiene,axole,rcra waste number u124,furfurane
8029
CHEBI:35559
furan
C1=COC=C1
110-00-9,128-37-0
C4H4O
68.06
MFCD00003222
YLQBMQCUIZJEEH-UHFFFAOYSA-N
divinylene oxide,furfuran,tetrole,oxole,oxacyclopentadiene,furane,1,4-epoxy-1,3-butadiene,axole,rcra waste number u124,furfurane
8029
CHEBI:35559
furan
C1=COC=C1
Water 0.2% max.
Refractive Index 1.4197 to 1.4217
Specific Gravity 0.936
Flash Point −35°C
Packaging Glass bottle
Density 0.9360g/mL
Color Colorless to Yellow
Melting Point -85.6°C
Quantity 250mL
Boiling Point 31.3°C
Infrared Spectrum Authentic
Stabilizer 0.025 to 0.04% BHT
Assay Percent Range 0.05% max. (Furfural) (GC)
UN Number 2389
Beilstein 17, 27
Fieser 07,161; 09,227; 10,187; 13,173
Merck Index 15, 4325
Solubility Information Solubility in water: insoluble. Other solubilities: freely soluble in alcohol,ether and aceton,hydrolysis in water
Formula Weight 68.06
Physical Form Liquid
Percent Purity 99+%
Chemical Name or Material Furan
Show More Show Less

GHS H Statement
Suspected of causing genetic defects.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Harmful if inhaled.
May cause cancer.
Harmful to aquatic life with long

GHS P Statement
Use personal protective equipment as required.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the

GHS Signal Word: Danger

EINECSNumber : 203-727-3