Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl Acetate 97.0+%, TCI America™
SDP

Catalog No. A0903100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0903100ML 100 mL
A090325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0903100ML Supplier TCI America Supplier No. A0903100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-17-6
Molecular Formula C7H8O3
Molecular Weight (g/mol) 140.138
MDL Number MFCD00003251
InChI Key CKOYRRWBOKMNRG-UHFFFAOYSA-N
Synonym furfuryl acetate, 2-furanmethanol, acetate, 2-furanmethanol acetate, 2-acetoxymethylfuran, furfuryl alcohol, acetate, acetic acid furfurylester, acetic acid furfuryl ester, 2-furanmethyl acetate, 2-furfuryl acetate, 2-furylmethyl acetate
PubChem CID 12170
IUPAC Name furan-2-ylmethyl acetate
SMILES CC(=O)OCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 177°C
Quantity 100 mL
Formula Weight 140.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Acetate
EINECSNumber (5)-0032
RTECSNumber LU9120000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.