Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl alcohol, ≥97.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018275
Click to view available options
Quantity:
1 mL
5 mL

Furfuryl alcohol belongs to the furan class of compounds.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 915°F
CAS 98-00-0
Vapor Pressure 0.5 mmHg (20°C)
Vapor Density 3.4 (vs. air)
Density 1.135 g/mL (at 25°C)
Flash Point 65°C - Closed Cup
Physical Form Neat
Linear Formula C5H6O2
MDL Number MFCD00003252
Melting Point -29°C
Boiling Point 170°C
Synonym 2-(Hydroxymethyl)furan
Type Analytical Standard
Quantity 5 mL
Formula Weight 98.10
Percent Purity ≥97.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.