Learn More
Furfuryl alcohol, 98%
Description
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Furfuryl alcohol was used in the synthesis of continuous titanium carbide nanofibers/nanoribbon. Furfuryl alcohol is an efficient trapping agent for singlet oxygen determination in natural waters.
Solubility
Soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep containers tightly closed in a cool, well-ventilated place. Keep in properly labeled containers. Store at room temperature. Incompatible with strong oxidizing agents.

Specifications
Specifications
Melting Point | -31°C |
Density | 1.129 |
Boiling Point | 170°C to 171°C |
Flash Point | 65°C (149°F) |
Odor | Mild |
Refractive Index | 1.487 |
Quantity | 1000 g |
UN Number | UN2874 |
Beilstein | 106291 |
Merck Index | 14,4305 |
Show More |
Safety and Handling
Hazard Category | H227-H301+H311-H319-H330-H335-H373 |
Hazard Statement | GHS H Statement H331-H351-H373-H227-H302-H312-H319-H335 Toxic if inhaled. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Combustible liquid. Harmful if swallowed. Harmful in contact with skin. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P262-P264-P264b-P270-P271-P280-P285-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P362-P374-P501c |
DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: FURFURYL ALCOHOL |
EINECSNumber | 202-626-1 |
RTECSNumber | LU9100000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.