Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl glycidyl ether, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
5380-87-0
C8H10O3
154.17
MFCD00013348
RUGWIVARLJMKDM-UHFFFAOYSA-N
furfuryl glycidyl ether,2-oxiran-2-ylmethoxy methyl furan,2,3-epoxypropyl-furfurylether,2-oxiranylmethoxy methyl furan,2-2,3-epoxypropoxy methyl furan,furan, 2-oxiranylmethoxy methyl,furan, 2-2,3-epoxypropoxy methyl
94306
2-(oxiran-2-ylmethoxymethyl)furan
C1C(O1)COCC2=CC=CO2
5380-87-0
C8H10O3
154.17
MFCD00013348
RUGWIVARLJMKDM-UHFFFAOYSA-N
furfuryl glycidyl ether,2-oxiran-2-ylmethoxy methyl furan,2,3-epoxypropyl-furfurylether,2-oxiranylmethoxy methyl furan,2-2,3-epoxypropoxy methyl furan,furan, 2-oxiranylmethoxy methyl,furan, 2-2,3-epoxypropoxy methyl
94306
2-(oxiran-2-ylmethoxymethyl)furan
C1C(O1)COCC2=CC=CO2
Specific Gravity 1.122
Density 1.1220g/mL
Color Colorless to Orange
Boiling Point 103.0°C to 104.0°C (11.0 mmHg)
Refractive Index 1.4800 to 1.4820
Quantity 5g
Packaging Glass bottle
Infrared Spectrum Authentic
Flash Point 102°C
Assay Percent Range 96% min. (GC)
Formula Weight 154.17
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Furfuryl glycidyl ether
Show More Show Less

GHS H Statement:
Harmful if inhaled.
May form explosive peroxides.

GHS P Statement:
Keep container tightly closed.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-372-6

RTECSNumber : LU1423000