Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl mercaptan, 98+%

Catalog No. AAB2238022
Click to view available options
Quantity:
25 g
100 g

CAS: 98-02-2 | C5H6OS | 114.162 g/mol

Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Furfuryl Mercaptam is a volatile flavor component of corn tortilla chips.

Solubility
Insoluble in water.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98-02-2
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.162
MDL Number MFCD00003254
InChI Key ZFFTZDQKIXPDAF-UHFFFAOYSA-N
Synonym furfuryl mercaptan, 2-furanmethanethiol, 2-furylmethanethiol, furfuryl thiol, 2-furylmethyl mercaptan, 2-furfurylmercaptan, furan-2-yl-methanethiol, 2-mercaptomethyl furan, furfurylmercaptan, 2-furfurylthiol
PubChem CID 7363
IUPAC Name furan-2-ylmethanethiol
SMILES C1=COC(=C1)CS

Specifications

Density 1.132
Boiling Point 155°C
Flash Point 45°C (113°F)
Odor Strong
Refractive Index 1.5315
Quantity 100 g
UN Number UN1228
Beilstein 383594
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 114.17
Percent Purity ≥98%
Chemical Name or Material Furfuryl mercaptan
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H331-H226-H302-H312-H319-H335
Toxic if inhaled.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber 202-628-2
RTECSNumber LU2100000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.