Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl methyl sulfide, 97%, Thermo Scientific™

Catalog No. AC310520250
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 1438-91-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00009604
InChI Key SKSFHXVDHVKIBN-UHFFFAOYSA-N
Synonym furfuryl methyl sulfide, methyl furfuryl sulfide, 2-methylthio methyl furan, 2-furfuryl methyl sulfide, furan, 2-methylthio methyl, 2-methylsulfanylmethyl furan, unii-lmb8355w32, 2-methylsulfanyl methyl furan, 2-methylthio methylfuran
PubChem CID 518937
IUPAC Name 2-[(methylsulfanyl)methyl]furan
SMILES CSCC1=CC=CO1

Specifications

Color Brown or Green to Yellow
Density 1.0700g/mL
Boiling Point 64.0°C to 65.0°C (15.0 mmHg)
Flash Point 63°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Refractive Index 1.5200 to 1.5260
Quantity 25 g
Formula Weight 128.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Furfuryl methyl sulfide, 97%
Show More Show Less
Hazard Category STENCH
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Product may darken on storage

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.