Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfurylamine, 99%

Catalog No. AAB2397536
Click to view available options
Quantity:
100 g
500 g

Catalog No. AAB2397536

Supplier: Thermo Scientific Chemicals B2397536

Only null left
Add to Cart

CAS: 617-89-0 | C5H7NO | 97.117 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Furfurylamine is used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics, antihypertensive, and antiseptic agents.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are strong oxidizing agents, air, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-89-0
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.117
MDL Number MFCD00003258
InChI Key DDRPCXLAQZKBJP-UHFFFAOYSA-N
Synonym furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
PubChem CID 3438
IUPAC Name furan-2-ylmethanamine
SMILES C1=COC(=C1)CN

Specifications

Melting Point -70°C
Density 1.05
pH 11.6
Boiling Point 145°C to 146°C
Flash Point 37°C (99°F)
Odor Amine-like
Refractive Index 1.49
Quantity 500 g
UN Number UN2526
Beilstein 1614
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 97.12
Percent Purity 99%
Chemical Name or Material Furfurylamine
Show More Show Less
Hazard Category H226-H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318-H226-H302-H312-H332
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FURFURYLAMINE
EINECSNumber 210-536-9
RTECSNumber LU9275000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.