Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfurylamine, 99+%

Catalog No. p-137549
Change view
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC119800050 5 mL Glass bottle
AC119802500 250 mL Glass bottle
AC119800010 1 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC119800050 Supplier Thermo Scientific Chemicals Supplier No. 119800050
Only null left
Add to Cart

CAS: 617-89-0 | C5H7NO | 97.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-89-0
Molecular Formula C5H7NO
Molecular Weight (g/mol) 97.12
MDL Number MFCD00003258
InChI Key DDRPCXLAQZKBJP-UHFFFAOYSA-N
Synonym furfurylamine, 2-furfurylamine, 2-furanmethylamine, 2-aminomethylfuran, 2-aminomethyl furan, 2-furanmethanamine, 2-furylmethylamine, 2-furylmethyl amine, usaf q-1, methylamine, 1-2-furyl
PubChem CID 3438
IUPAC Name furan-2-ylmethanamine
SMILES C1=COC(=C1)CN

Specifications

Melting Point -70°C
Color Brown-Yellow to Colorless
Density 1.0900g/mL
Boiling Point 145°C to 146°C (761.0 mmHg)
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Glass bottle
Refractive Index 1.4890 to 1.491
Quantity 5 mL
Beilstein 18, 584
Specific Gravity 1.09
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether
Viscosity 1.6 mPa.s (20°C)
Formula Weight 97.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Furfurylamine, 99+%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove victim to fresh air and
EINECSNumber 210-536-9
RTECSNumber LU9275000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.