Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FURIN (Human) ELISA Kit

Catalog No. p-4392790 Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit
96 Assays
Sample Type:
CCS, Cell Lysates, Tissue Lysates
CCS, Plasma, Serum, Urine

In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human FURIN.

Specifications

Calibration Range 312 to 20000pg/mL
Cross Reactivity Human
Description FURIN (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human FURIN.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Cell Lysates, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type FURIN (Human) ELISA Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.