Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furin Mouse anti-Human, Unconjugated, Clone: 16B1.1, MilliporeSigma™

Catalog No. MABS181225 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1812-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS1812-25 Supplier MilliporeSigma Supplier No. MABS181225UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Furin, clone 16B1.1, Cat. No. MABS1812, is a mouse monoclonal antibody that detects Furin in human cells and has been tested for use in Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Furin
Applications Western Blot
Classification Monoclonal
Clone 16B1.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2b in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias EC: 3.4.2.75;Dibasic-processing enzyme;Paired basic amino acid residue-cleaving enzyme;PACE
Gene Symbols FURIN;FUR;PACE;PCSK3
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 177 amino acids from the C-terminal half of human Furin.
Molecular Weight of Antigen ∽100 kDa observed; 86.68 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Test Specificity Clone 16B1.1 detects human Furin and it targets an epitope within 177 amino acids from the C-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Product Type Antibodies
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.