Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furosemide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602878
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle

F1133, 54-31-9, C12H11ClN2O5S

Spectrum™ Chemical Furosemide, USP is used to reduce extra fluid in the body (edema) caused by conditions such as heart failure, liver disease, and kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12H11ClN2O5S
Molecular Weight (g/mol) 330.74
InChI Key ZZUFCTLCJUWOSV-UHFFFAOYSA-N
IUPAC Name 4-chloro-2-{[(furan-2-yl)methyl]amino}-5-sulfamoylbenzoic acid
SMILES NS(=O)(=O)C1=CC(C(O)=O)=C(NCC2=CC=CO2)C=C1Cl

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.