Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FUS Antibody, DyLight 680, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100560FR
Description
FUS Polyclonal antibody specifically detects FUS in Human, Mouse samples. It is validated for Western BlotSpecifications
FUS | |
Polyclonal | |
Western Blot | |
75 kDa DNA-pairing protein, ALS6, amyotrophic lateral sclerosis 6, FUS1, fused in sarcoma, fusion (involved in t(12;16) in malignant liposarcoma), fusion gene in myxoid liposarcoma, fusion, derived from t(12;16) malignant liposarcoma, fus-like protein, heterogeneous nuclear ribonucleoprotein P2, HNRNPP2, Oncogene FUS, Oncogene TLS, POMp75, RNA-binding protein FUS, TLShnRNP-P2, Translocated in liposarcoma protein | |
A synthetic peptide which maps to a region between residues 200 and 250 of human fusion (involved in t(12;16) in malignant liposarcoma) using the numbering given in SwissProt entry P35637 (GeneID 2521). | |
0.1 mL | |
Cancer, Neuroscience | |
2521 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction